Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAMPTP1
Entrez Gene ID
417707
Gene Name
nicotinamide phosphoribosyltransferase pseudogene 1
Gene Aliases
NAMPT, PBEF1, visfatin
Chromosome Location
Chr.1: 14363077 - 14392439 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030728.1 NP_001025899.1 3 - 4 377 103
XM_015275716.2 XP_015131202.1 3 - 4 630 103
GenBank mRNA AJ719830.1 - 3 - 4 417 103
AY946242.1 - 3 - 4 377 103

View Full Product Details