Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB9
Entrez Gene ID
417783
Gene Name
DnaJ heat shock protein family (Hsp40) member B9
Gene Aliases
-
Chromosome Location
Chr.1: 28562682 - 28571177 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030735.1 NP_001025906.1 2 - 3 275 126
GenBank mRNA AJ720657.1 - 2 - 3 275 126

View Full Product Details