Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM60A
Entrez Gene ID
418140
Gene Name
family with sequence similarity 60 member A
Gene Aliases
SINHCAF
Chromosome Location
Chr.1: 59546382 - 59561674 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030766.1 NP_001025937.1 4 - 5 596 102
XM_015285715.2 XP_015141201.1 4 - 5 648 102
XM_015285722.2 XP_015141208.1 4 - 5 409 102
XM_025148812.1 XP_025004580.1 4 - 5 633 102
GenBank mRNA AJ720327.1 - 4 - 5 596 102

View Full Product Details