Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OVALY
Entrez Gene ID
420897
Gene Name
ovalbumin-related protein Y (SERPINB14B)
Gene Aliases
SERPINB14B, SERPINB3, oval-Y
Chromosome Location
Chr.2: 67700179 - 67707207 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031001.1 NP_001026172.1 3 - 4 284 87
XM_015282171.1 XP_015137657.1 2 - 3 2243 87
GenBank mRNA AB288388.1 - 2 - 3 220 87
AB288389.1 - 2 - 3 220 87
AB288390.1 - 2 - 3 220 87

View Full Product Details