Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DAP
Entrez Gene ID
420923
Gene Name
death associated protein
Gene Aliases
DAP1
Chromosome Location
Chr.2: 78125292 - 78169245 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031003.1 NP_001026174.1 2 - 3 195 102
GenBank mRNA AJ720932.1 - 2 - 3 195 102
JQ268269.1 - 2 - 3 164 102

View Full Product Details