Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGCB
Entrez Gene ID
422760
Gene Name
sarcoglycan beta
Gene Aliases
beta-sarcoglycan
Chromosome Location
Chr.4: 66162127 - 66168332 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031155.1 NP_001026326.1 3 - 4 446 77
XM_015285438.2 XP_015140924.1 3 - 4 445 77
XM_015285439.2 XP_015140925.1 3 - 4 686 77
GenBank mRNA AJ720367.1 - 3 - 4 446 77

View Full Product Details