Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDFY1
Entrez Gene ID
424806
Gene Name
WD repeat and FYVE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 8445709 - 8469090 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031310.1 NP_001026481.1 10 - 11 1089 122
GenBank mRNA AJ720946.1 - 10 - 11 1089 122

View Full Product Details