Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHO1
Entrez Gene ID
426108
Gene Name
pleckstrin homology domain containing O1
Gene Aliases
-
Chromosome Location
Chr.25: 115449 - 131029 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031356.1 NP_001026527.1 3 - 4 585 72
XM_025143406.1 XP_024999174.1 3 - 4 531 72
XM_025143407.1 XP_024999175.1 3 - 4 522 72
GenBank mRNA AJ851722.1 - 3 - 4 585 72
KY982274.1 - 3 - 4 350 72

View Full Product Details