Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL28
Entrez Gene ID
426121
Gene Name
mitochondrial ribosomal protein L28
Gene Aliases
-
Chromosome Location
Chr.14: 12534748 - 12537899 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031359.2 NP_001026530.2 2 - 3 467 75
GenBank mRNA BU311325.1 - 2 - 3 184 75
BU311826.1 - 1 - 2 95 75
BU380572.1 - 2 - 3 467 75
CO419869.1 - 3 - 4 663 75

View Full Product Details