Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNSTRN
Entrez Gene ID
426704
Gene Name
kinetochore-localized astrin/SPAG5 binding protein
Gene Aliases
C15ORF23, C5H15orf23, HSD11, SKAP, TRAF4AF1, kinastrin
Chromosome Location
Chr.5: 1014712 - 1019729 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031393.1 NP_001026564.1 4 - 5 440 74
GenBank mRNA AJ721023.1 - 4 - 5 440 74
CR352610.1 - 3 - 4 294 74

View Full Product Details