Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHD1
Entrez Gene ID
424741
Gene Name
EF-hand domain family member D1
Gene Aliases
-
Chromosome Location
Chr.9: 1130069 - 1145060 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031773.1 NP_001026943.1 1 - 2 396 84
GenBank mRNA AY971655.1 - 1 - 2 396 84
BX933977.2 - 1 - 2 299 84

View Full Product Details