Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CETP
Entrez Gene ID
415645
Gene Name
cholesteryl ester transfer protein
Gene Aliases
-
Chromosome Location
Chr.11: 615984 - 629273 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034814.2 NP_001029986.2 14 - 15 1380 82
XM_025154152.1 XP_025009920.1 21 - 22 5664 82
GenBank mRNA AB205527.1 - 14 - 15 1406 82

View Full Product Details