Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XKR9
Entrez Gene ID
428363
Gene Name
XK related 9
Gene Aliases
XRG9
Chromosome Location
Chr.2: 116771933 - 116782420 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034824.1 NP_001029996.1 1 - 2 311 116
XM_015282822.2 XP_015138308.2 3 - 4 2512 116
XM_015282823.2 XP_015138309.2 2 - 3 702 116
GenBank mRNA AY699714.1 - 1 - 2 311 116

View Full Product Details