Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT7B
Entrez Gene ID
427937
Gene Name
Wnt family member 7B
Gene Aliases
-
Chromosome Location
Chr.1: 70751533 - 70843213 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037274.1 NP_001032351.1 1 - 2 143 87
GenBank mRNA AY753290.1 - 1 - 2 143 87

View Full Product Details