Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMARCB1
Entrez Gene ID
769489
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1
Gene Aliases
MMP11
Chromosome Location
Chr.15: 8283579 - 8293534 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039255.1 NP_001034344.1 4 - 5 434 63
GenBank mRNA AJ720244.1 - 4 - 5 434 63
BX931383.1 - - 429 63
BX932666.2 - 4 - 5 430 63
KU325499.1 - 3 - 4 253 63
KU557484.1 - 3 - 4 253 63

View Full Product Details