Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAMP3
Entrez Gene ID
419368
Gene Name
vesicle-associated membrane protein 3
Gene Aliases
-
Chromosome Location
Chr.21: 310699 - 314464 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039489.1 NP_001034578.1 4 - 5 375 85
GenBank mRNA BI067069.1 - 2 - 3 90 85
BM488089.1 - 4 - 5 377 85
BX929338.2 - 4 - 5 363 85

View Full Product Details