Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPR
Entrez Gene ID
419570
Gene Name
heterogeneous nuclear ribonucleoprotein R
Gene Aliases
HNRPR
Chromosome Location
Chr.23: 1662494 - 1685588 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006309.3 NP_001006309.3 7 - 8 922 158
XM_015297543.2 XP_015153029.1 7 - 8 972 158
XM_015297545.2 XP_015153031.1 5 - 6 672 158
XM_015297546.2 XP_015153032.1 6 - 7 742 158
XM_025142983.1 XP_024998751.1 7 - 8 913 158
GenBank mRNA AJ446673.1 - 2 - 3 304 158
AJ720877.1 - 7 - 8 917 158
CN779270.1 - 4 - 5 533 158

View Full Product Details