Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
N4BP2L2
Entrez Gene ID
418912
Gene Name
NEDD4 binding protein 2 like 2
Gene Aliases
PFAAP5
Chromosome Location
Chr.1: 175842295 - 175887181 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015277885.2 XP_015133371.1 5 - 6 2041 91
XM_015277889.2 XP_015133375.1 5 - 6 2023 91
XM_015277900.2 XP_015133386.1 5 - 6 2041 91
XM_015277908.2 XP_015133394.1 5 - 6 2039 91
XM_015277912.2 XP_015133398.1 5 - 6 2039 91
XM_025146830.1 XP_025002598.1 5 - 6 2038 91
GenBank mRNA CR386445.1 - 2 - 3 336 91
CR390564.1 - 4 - 5 427 91

View Full Product Details