Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC2L1
Entrez Gene ID
419406
Gene Name
cell division cycle 2-like 1 (PITSLRE proteins)
Gene Aliases
CDC2L2, CDK11A, cPITSLRE
Chromosome Location
Chr.21: 2016024 - 2032084 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030871.3 NP_001026042.2 16 - 17 1883 85
XM_015296863.2 XP_015152349.1 17 - 18 4733 85
XM_015296864.2 XP_015152350.1 17 - 18 4730 85
XM_015296865.2 XP_015152351.1 17 - 18 4727 85
XM_015296866.2 XP_015152352.1 16 - 17 4610 85
XM_025142506.1 XP_024998274.1 17 - 18 4751 85
XM_025142508.1 XP_024998276.1 16 - 17 4607 85

View Full Product Details