Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDSUB1
Entrez Gene ID
424331
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 36520588 - 36550929 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031263.2 NP_001026434.2 4 - 5 1009 111
XM_015290154.1 XP_015145640.1 3 - 4 1081 111
XM_015290155.1 XP_015145641.1 3 - 4 1081 111
XM_015290156.1 XP_015145642.1 3 - 4 1712 111
XM_015290160.2 XP_015145646.1 5 - 6 1168 111
XM_015290161.2 XP_015145647.1 5 - 6 1139 111
XM_015290162.2 XP_015145648.1 5 - 6 1313 111
XM_015290163.2 XP_015145649.1 4 - 5 1051 111
XM_015290164.2 XP_015145650.1 4 - 5 1026 111
XM_015290165.2 XP_015145651.1 4 - 5 786 111
XM_015290166.2 XP_015145652.1 5 - 6 935 111
XM_015290167.2 XP_015145653.1 4 - 5 578 111
XM_025152249.1 XP_025008017.1 3 - 4 1081 111
XM_025152250.1 XP_025008018.1 5 - 6 1277 111
XM_025152252.1 XP_025008020.1 5 - 6 1352 111
XM_025152253.1 XP_025008021.1 5 - 6 1281 111
XM_025152254.1 XP_025008022.1 5 - 6 1143 111
XM_025152255.1 XP_025008023.1 5 - 6 1193 111
XM_025152256.1 XP_025008024.1 5 - 6 903 111
XM_025152257.1 XP_025008025.1 5 - 6 1197 111
XM_025152258.1 XP_025008026.1 4 - 5 585 111
GenBank mRNA AJ719461.1 - 4 - 5 1014 111
AJ726210.1 - 3 - 4 479 111

View Full Product Details