Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NTF3
Entrez Gene ID
428099
Gene Name
neurotrophin 3
Gene Aliases
HDNF, NGF-2, NGF2, NT-3, NT3, neurotrophin-3
Chromosome Location
Chr.1: 74215247 - 74285493 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031488.1 NP_001026659.1 1 - 1 746 100
NM_001109762.1 NP_001103232.1 2 - 2 806 100

View Full Product Details