Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASIC1
Entrez Gene ID
426883
Gene Name
acid sensing ion channel subunit 1
Gene Aliases
ACCN2
Chromosome Location
Chr.33: 5796495 - 5814160 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040467.1 NP_001035557.1 10 - 11 1486 55
XM_015300232.2 XP_015155718.1 9 - 10 3960 55
XM_015300233.2 XP_015155719.1 9 - 10 2469 55
XM_015300234.2 XP_015155720.1 11 - 12 1960 55
XM_015300235.2 XP_015155721.1 11 - 12 1645 55
GenBank mRNA AY956393.1 - 10 - 11 1486 55
CR389178.1 - - 53 55

View Full Product Details