Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NSL1
Entrez Gene ID
421367
Gene Name
NSL1, MIS12 kinetochore complex component
Gene Aliases
-
Chromosome Location
Chr.3: 21899749 - 21912897 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044646.1 NP_001038111.1 5 - 6 591 107
XM_025148669.1 XP_025004437.1 5 - 6 456 107
GenBank mRNA AB253767.1 - 5 - 6 591 107

View Full Product Details