Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPY6R
Entrez Gene ID
693264
Gene Name
neuropeptide Y receptor Y6 (pseudogene)
Gene Aliases
Y6
Chromosome Location
Chr.13: 15166454 - 15174227 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044687.1 NP_001038152.1 1 - 1 1051 104
XM_015293952.2 XP_015149438.1 4 - 4 3465 104
GenBank mRNA DQ189216.1 - 1 - 1 1051 104

View Full Product Details