Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNRH1
Entrez Gene ID
770134
Gene Name
gonadotropin releasing hormone 1
Gene Aliases
CGNRH-I, GnRH-I, LHRH-I
Chromosome Location
Chr.22: 973244 - 976247 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080877.1 NP_001074346.1 2 - 3 197 77

View Full Product Details