Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TESC
Entrez Gene ID
771113
Gene Name
tescalcin
Gene Aliases
TSC
Chromosome Location
Chr.15: 11627661 - 11631427 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080884.2 NP_001074353.1 2 - 3 225 103
XM_025155510.1 XP_025011278.1 2 - 3 189 103
GenBank mRNA BX931037.1 - 2 - 3 226 103

View Full Product Details