Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
MRPL37
Entrez Gene ID
772313
Gene Name
mitochondrial ribosomal protein L37
Gene Aliases
-
Chromosome Location
Chr.8: 25514945 - 25520455 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081505.1 NP_001074974.1 2 - 3 587 84
GenBank mRNA AJ720915.1 - 2 - 3 587 84

View Full Product Details