The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081505.1 | NP_001074974.1 | 5 - 6 | 1053 | 81 |
GenBank mRNA | AJ720915.1 | - | 5 - 6 | 1053 | 81 |