Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUSP1
Entrez Gene ID
374192
Gene Name
dual specificity phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.13: 9770031 - 9772637 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085359.2 NP_001078828.1 1 - 2 471 78
GenBank mRNA AF026522.1 - 1 - 2 338 78
CR387287.1 - 1 - 2 257 78

View Full Product Details