Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYL10
Entrez Gene ID
417506
Gene Name
myosin, light chain 10, regulatory
Gene Aliases
G2, MLC-2B, MYLC2PL
Chromosome Location
Chr.19: 3899866 - 3931151 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097526.1 NP_001090995.1 2 - 3 126 96
XM_015295706.1 XP_015151192.1 3 - 4 387 96
GenBank mRNA CR353595.1 - 3 - 4 391 96
EF463065.1 - 2 - 3 126 96

View Full Product Details