Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM7.2
Entrez Gene ID
425772
Gene Name
tripartite motif containing 7.2
Gene Aliases
TRIM27
Chromosome Location
Chr.16: 2454629 - 2458853 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099354.1 NP_001092824.1 3 - 4 817 108

View Full Product Details