The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112662.1 | NP_001106132.1 | 6 - 7 | 504 | 68 |
XM_015293764.2 | XP_015149250.1 | 7 - 8 | 648 | 68 | |
GenBank mRNA | BX260174.3 | - | 2 - 3 | 158 | 68 |
BX277920.4 | - | 6 - 7 | 504 | 68 |