Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX5
Entrez Gene ID
373988
Gene Name
T-box 5
Gene Aliases
-
Chromosome Location
Chr.15: 12598201 - 12638588 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204173.1 NP_989504.1 4 - 5 511 70
XM_015294830.2 XP_015150316.1 3 - 4 1055 70
XM_015294831.2 XP_015150317.1 4 - 5 1239 70
XM_015294832.1 XP_015150318.1 4 - 5 466 70
GenBank mRNA AF033671.1 - 4 - 5 450 70
AF069396.2 - 4 - 5 511 70
CR353590.1 - 4 - 5 576 70

View Full Product Details