Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENTPD8
Entrez Gene ID
374095
Gene Name
ectonucleoside triphosphate diphosphohydrolase 8
Gene Aliases
ENTPD1
Chromosome Location
Chr.17: 1280057 - 1290211 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204247.2 NP_989578.2 9 - 10 1623 66
XM_015279254.2 XP_015134740.1 9 - 10 3609 66
XM_015279258.2 XP_015134744.1 9 - 10 3622 66
XM_025156014.1 XP_025011782.1 9 - 10 3418 66
XM_025156015.1 XP_025011783.1 9 - 10 3406 66
XM_025156016.1 XP_025011784.1 8 - 9 4575 66
XM_025156017.1 XP_025011785.1 9 - 10 2279 66
GenBank mRNA AF041355.1 - 8 - 9 66
AF426405.1 - 8 - 9 1294 66

View Full Product Details