Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNAI3
Entrez Gene ID
374097
Gene Name
G protein subunit alpha i3
Gene Aliases
Galphai3
Chromosome Location
Chr.26: 1245972 - 1252794 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204249.1 NP_989580.1 - 880 144
GenBank mRNA L24551.1 - - 880 144

View Full Product Details