Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSHB
Entrez Gene ID
374108
Gene Name
follicle stimulating hormone beta subunit
Gene Aliases
-
Chromosome Location
Chr.5: 4772499 - 4775874 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204257.1 NP_989588.1 2 - 3 203 71
GenBank mRNA AB077362.1 - 1 - 2 159 71
AY029204.1 - 2 - 3 203 71
GQ856365.1 - 1 - 2 159 71

View Full Product Details