Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT7A
Entrez Gene ID
374168
Gene Name
Wnt family member 7A
Gene Aliases
-
Chromosome Location
Chr.12: 6435529 - 6467522 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204292.1 NP_989623.1 2 - 3 372 57
GenBank mRNA AB045629.1 - 2 - 3 372 57

View Full Product Details