Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCX
Entrez Gene ID
374242
Gene Name
doublecortin
Gene Aliases
18C15.5
Chromosome Location
Chr.4: 13185758 - 13314342 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001305674.2 NP_001292603.1 3 - 4 893 76
NM_204335.3 NP_989666.1 3 - 4 893 76
XM_015278173.2 XP_015133659.1 6 - 7 3375 76
XM_015278174.2 XP_015133660.1 5 - 6 1066 76
XM_015278175.2 XP_015133661.1 4 - 5 1058 76
XM_015278176.2 XP_015133662.1 4 - 5 971 76
XM_015278177.2 XP_015133663.1 6 - 7 3374 76
XM_015278178.2 XP_015133664.1 6 - 7 3374 76
XM_015278179.2 XP_015133665.1 6 - 7 3373 76
GenBank mRNA AF330009.1 - 3 - 4 779 76

View Full Product Details