Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFCP2
Entrez Gene ID
378900
Gene Name
transcription factor CP2
Gene Aliases
TCFCP2
Chromosome Location
Chr.33: 4984491 - 4998043 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204384.1 NP_989715.1 2 - 3 276 63
XM_015273097.2 XP_015128583.1 2 - 3 980 63
GenBank mRNA AB004542.1 - 2 - 3 326 63
AY298725.1 - 2 - 3 276 63

View Full Product Details