Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZP3
Entrez Gene ID
378906
Gene Name
zona pellucida sperm-binding protein 3
Gene Aliases
ZPC, Zp-3
Chromosome Location
Chr.10: 3087977 - 3090609 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204389.2 NP_989720.3 7 - 8 1118 80
GenBank mRNA BX258818.3 - 1 - 2 94 80
D89097.2 - 7 - 8 1118 80

View Full Product Details