Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
RHOQ
Entrez Gene ID
395114
Gene Name
ras homolog family member Q
Gene Aliases
CHTC10
Chromosome Location
Chr.3: 26838990 - 26854605 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204461.1 NP_989792.1 1 - 1 116 137
GenBank mRNA AF372468.1 - 1 - 1 116 137

View Full Product Details