Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPTC
Entrez Gene ID
395130
Gene Name
opticin
Gene Aliases
-
Chromosome Location
Chr.26: 5176525 - 5179970 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204473.1 NP_989804.1 - 503 86
XM_015298880.2 XP_015154366.1 - 484 86
XM_015298881.2 XP_015154367.1 - 450 86
GenBank mRNA AY156089.1 - - 503 86
BX935704.1 - - 464 86
CR405737.1 - - 421 86

View Full Product Details