Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BSX
Entrez Gene ID
395183
Gene Name
brain specific homeobox
Gene Aliases
-
Chromosome Location
Chr.24: 3127754 - 3129687 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204512.1 NP_989843.1 1 - 2 344 63
GenBank mRNA AY500594.1 - 1 - 2 344 63

View Full Product Details