Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MATN3
Entrez Gene ID
395954
Gene Name
matrilin 3
Gene Aliases
matrilin-3
Chromosome Location
Chr.3: 101394465 - 101409742 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205072.1 NP_990403.1 1 - 2 154 111
GenBank mRNA AJ000055.1 - 1 - 2 154 111

View Full Product Details