Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSN
Entrez Gene ID
395955
Gene Name
translin
Gene Aliases
C3PO
Chromosome Location
Chr.7: 26247175 - 26251594 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205073.1 NP_990404.1 4 - 5 372 82
GenBank mRNA X95074.1 - - 372 82

View Full Product Details