Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FTH1
Entrez Gene ID
395970
Gene Name
ferritin heavy chain 1
Gene Aliases
FTH
Chromosome Location
Chr.5: 16995949 - 17000513 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205086.1 NP_990417.1 3 - 4 531 67
GenBank mRNA BX934496.1 - 3 - 4 429 67
CR388929.1 - 2 - 3 275 67
Y14698.1 - - 531 67

View Full Product Details