Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GFRA2
Entrez Gene ID
395993
Gene Name
GDNF family receptor alpha 2
Gene Aliases
NTNRALPHA
Chromosome Location
Chr.22: 1714010 - 1734922 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205101.1 NP_990432.1 5 - 6 934 105
GenBank mRNA U90542.1 - 5 - 6 934 105

View Full Product Details