Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BFSP1
Entrez Gene ID
396056
Gene Name
beaded filament structural protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 12061525 - 12107620 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205151.1 NP_990482.1 6 - 7 1178 78
XM_025148550.1 XP_025004318.1 13 - 14 3717 78
XM_025148551.1 XP_025004319.1 12 - 13 3607 78
GenBank mRNA X72873.1 - - 1178 78

View Full Product Details