Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYAB
Entrez Gene ID
396089
Gene Name
crystallin alpha B
Gene Aliases
alpha(B)-crystallin
Chromosome Location
Chr.24: 6242317 - 6246235 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205176.1 NP_990507.1 2 - 3 334 153
GenBank mRNA BX930014.2 - 2 - 3 349 153
S53164.1 - 2 - 3 334 153
U26661.1 - 2 - 3 514 153

View Full Product Details