Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA9
Entrez Gene ID
415540
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C10H15ORF44, C15orf44, INTS14
Chromosome Location
Chr.10: 18494771 - 18505021 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005825.1 NP_001005825.1 5 - 6 665 131
XM_015292093.2 XP_015147579.1 5 - 6 742 131
GenBank mRNA AJ720263.1 - 5 - 6 665 131

View Full Product Details